Combustion Trays, Aluminium Oxide AL24, Thermo Scientific
Supplier: Thermo Scientific
33187-KF 33187-KF
CAAA33187-KFEA 622.52 CAD
AA33187-KF CAAA33187-KF
Combustion Trays, Aluminium Oxide AL24, Thermo Scientific
Boats Combustion Boats
Pure Al2O3 Aluminium oxide AL24.
Slightly porous, good resistance to thermal shock, extremely good creep strength.