Combustion Trays, Aluminium Oxide AL23, Thermo Scientific
Supplier: Thermo Scientific Chemicals
33183-KF 33184-KF 33185-KF 33186-KF
CAAA33183-KFEA 127.03 CAD
CAAA33183-KF CAAA33184-KF CAAA33185-KF CAAA33186-KF
Combustion Trays, Aluminium Oxide AL23, Thermo Scientific
Boats Combustion Boats
Pure Al2O3 Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.