Ceramic Plates, Rectangular/Square, AL25, Thermo Scientific
Supplier: Thermo Scientific Chemicals
33159-KF 33160-KF 33161-KF
CAAA33159-KFEA 279.66 CAD
CAAA33159-KF CAAA33160-KF CAAA33161-KF CAAA33162-KF
Ceramic Plates, Rectangular/Square, AL25, Thermo Scientific
Trays/Dishes/Pots Laboratory Trays/Dishes
Pure Al2O3 Aluminium oxide, AL25.
Very porous, good thermal insulation, highest resistance to thermal shock of all the Al2O3 materials