Ceramic Plates, Rectangular/Square, AL23, Thermo Scientific
Supplier: Thermo Scientific Chemicals
33155-KF 33156-KF 33158-KF
CAAA33155-KFEA 98.75 CAD
CAAA33155-KF CAAA33156-KF CAAA33158-KF
Ceramic Plates, Rectangular/Square, AL23, Thermo Scientific
Trays/Dishes/Pots Laboratory Trays/Dishes
Pure Al2O3 Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.