Ceramic Discs, Round, AL23, with Hole, Thermo Scientific
Supplier: Thermo Scientific
33143-KF 33146-KF 33144-KF 33142-KF 33140-KF 33146-KF 33147-KF 33142-KF 33144-KF 33147-KF 33140-KF 33143-KF
CAAA33146-KFEA 64.95 CAD
CAAA33146-KF CAAA33147-KF CAAA33144-KF AA33144-KF AA33146-KF AA33147-KF CAAA33142-KF CAAA33143-KF AA33142-KF AA33143-KF CAAA33140-KF AA33140-KF
Ceramic Discs, Round, AL23, with Hole, Thermo Scientific
Trays/Dishes/Pots Laboratory Trays/Dishes
Pure Al2O3 Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.