Fish Spine Beads, AL23, Thermo Scientific
Supplier: Thermo Scientific Chemicals
32905-KX 32906-KX 32907-KX 32908-KX
CAAA32905-KXEA 262.28 CAD
CAAA32905-KX CAAA32906-KX CAAA32907-KX CAAA32908-KX CAAA32909-KX
Fish Spine Beads, AL23, Thermo Scientific
Beads Ceramic Beads
Pure Al2O3 Aluminium oxide, AL 23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Order Now
SPECIFICATIONS
Environmentally Preferable