6-O-Palmitoyl-L-ascorbic acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: L-Ascorbic acid 6-palmitate, Palmitoyl-L-ascorbic acid, Ascorbyl Palmitate, (S)-2-((R)-3,4-Dihydroxy-5-oxo-2,5-dihydrofuran-2-yl)-2-hydroxyethyl palmitate, L(+)-Ascorbyl palmitate

32791-14 32791-22 32791-36
CAAA32791-14EA 61.3 CAD
CAAA32791-14 CAAA32791-22 CAAA32791-36
6-O-Palmitoyl-L-ascorbic acid 99%
6-O-Palmitoyl-L-ascorbic acid

Purity: 99% CAS No.: 137-66-6 Molecular Formula: C22H38O7 Formula Weight: 414.52 Physical Form: Crystalline Odor: Pleasant MDL No.: MFCD00005377

Formula: C₂₂H₃₈O₇
Boiling Pt: ∼250 °C
Melting Pt: 113…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00005377
CAS Number: 137-66-6
EINECS: 205-305-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR