Boron phosphate, (max. 10% H₂O)

Supplier: Thermo Scientific Chemicals
Warning

32562-22 32562-36
CAAA32562-22EA 237 CAD
CAAA32562-22 CAAA32562-36
Boron phosphate, (max. 10% H₂O)
Boron phosphate

H2O10%


CAS No.: 13308-51-5 Molecular Formula: BPO4 Formula Weight: 105.79 Physical Form: Powder Density: 2.81 Appearance: White Odor: Odorless Melting Point: 1400° MDL No.: MFCD00011318

Formula: BO₄P
MW: 105.78 g/mol
Melting Pt: >1400 °C
Density: 2.81
Storage Temperature: Ambient
MDL Number: MFCD00011318
CAS Number: 13308-51-5
EINECS: 236-337-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR