Ceramic Multibore Insulating Tubes, AL23, Round, Thermo Scientific
Supplier: Thermo Scientific Chemicals
32554-EW 32556-DV 32558-EL 32560-DZ
CAAA32550-DSEA 48.68 CAD
CAAA32550-DS CAAA32550-DV CAAA32550-DZ CAAA32550-EG CAAA32550-EL CAAA32550-ER CAAA32550-EW CAAA32550-FB CAAA32552-DS CAAA32552-DV CAAA32552-DZ CAAA32552-EG CAAA32552-EL CAAA32552-ER CAAA32552-EW CAAA32552-FB CAAA32554-DS CAAA32554-DV CAAA32554-DZ CAAA32554-EG CAAA32554-EL CAAA32554-ER CAAA32554-EW CAAA32554-FB CAAA32556-DS CAAA32556-DV CAAA32556-DZ CAAA32556-EG CAAA32556-EL CAAA32556-ER CAAA32556-EW CAAA32556-FB CAAA32558-DS CAAA32558-DV CAAA32558-DZ CAAA32558-EG CAAA32558-EL CAAA32558-ER CAAA32558-EW CAAA32558-FB CAAA32560-DS CAAA32560-DV CAAA32560-DZ CAAA32560-EG CAAA32560-EL CAAA32560-ER CAAA32560-EW CAAA32560-FB
Ceramic Multibore Insulating Tubes, AL23, Round, Thermo Scientific
Tubing Transfer Tubing
Pure Al₂O₃ Aluminium oxide, AL23.
  • Corrosion resistant
  • Permeable to microwaves
Dense, excellent thermal and electrical resistance properties.