Zinc protoporphyrin ≥96%

Supplier: Thermo Scientific Chemicals

Synonyms: Zinc(II) 3,3'-(3,8,13,17-tetramethyl-7,12-divinylporphyrin-2,18-diyl)dipropionate, Protoporphyrin IX zinc(II), Zinc (II) Protoporphyrin IX

32068-01 32068-MA 32068-MF
CAAAA32068-01EA 823.18 CAD
CAAAA32068-01 CAAA32068-MA CAAA32068-MF
Zinc protoporphyrin ≥96%
Zinc protoporphyrin

Purity: 96% CAS No.: 15442-64-5 Molecular Formula: C34H32N4O4Zn Formula Weight: 626.04 Physical Form: Crystalline Appearance: Dark red to purple Odor: Odorless MDL No.: MFCD00011612

Formula: C₃₄H₃₂N₄O₄Zn
MW: 626.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011612
CAS Number: 15442-64-5
EINECS: 239-455-7

Order Now

Specification Test Results

Form Crystalline


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR