Mineral oil, white (petroleum)
Supplier: Thermo Scientific Chemicals
Synonyms:
Matrix oil, Mineral oil, Paraffin oil
31911-A1
31911-D9
CAAA31911-A1EA
65.89
CAD
CAAA31911-A1
CAAA31911-D9
Mineral oil, white (petroleum)
White mineral oil (petroleum)
Sp. Gr. 0.849-0.864; Viscosity at 40°: 23.9-26.3 cSt
CAS No.: 8020-83-5
Physical Form: Liquid
Appearance: Colorless
Odor: Mineral-oil-like
MDL No.: MFCD00131611
Flash Pt:
>100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00131611 CAS Number: 8042-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...