Tripropylene glycol (mixture of isomers)
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Tripropylene glycol
31798-AK
31798-K2
31798-K7
CAAA31798-AKEA
46.29
CAD
CAAA31798-AK
CAAA31798-K2
CAAA31798-K7
Tripropylene glycol (mixture of isomers)
Tripropylene glycol (mixture of isomers)
CAS No.: 24800-44-0
Molecular Formula: HOCH(CH3)CH2OCH2CH(CH3)OCH2CH(CH3)OH
Formula Weight: 192.25
Physical Form: Liquid
Density: 1.018
Refractive Index: 1.4443
Odor: Odorless
Boiling Point: 268°
MDL No.: MFCD00014405
Formula:
C₉H₂₀O₄ MW: 192.26 g/mol Boiling Pt: 268 °C Density: 1.018 Storage Temperature: Ambient |
MDL Number:
MFCD00014405 CAS Number: 24800-44-0 EINECS: 246-466-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...