Ceramic Insulating Tubes, AL23, Both Ends Open, Thin Wall, Thermo Scientific
Supplier: Thermo Scientific Chemicals
31475-DM 31475-DR 31475-DT 31475-DW 31476-DR 31476-DT 31477-DW 31478-DM 31478-DR 31478-DW 31479-DM 31479-DR 31479-DW 31482-DT 31482-DW 31485-DR 31485-DW 31486-DM 31486-DR 31487-DR 31487-DW 31488-DM 31488-DW 31489-DM 31490-NP 31492-DM 31492-DR 031484-QAT 031485-QAT 031486-QAT 031488-QAT 031489-QAT 031490-QAT 031492-QAT
CAAA31475-DMEA 35.71 CAD
CAAA31475-DM CAAA31475-DR CAAA31475-DT CAAA31475-DW CAAA31476-DM CAAA31476-DR CAAA31476-DT CAAA31476-DW CAAA31477-DM CAAA31477-DR CAAA31477-DW CAAA31478-DM CAAA31478-DR CAAA31478-DW CAAA31479-DM CAAA31479-DR CAAA31479-DW CAAA31480-DM CAAA31480-DR CAAA31480-DW CAAA31482-DM CAAA31482-DR CAAA31482-DT CAAA31482-DW CAAA31484-DM CAAA31484-DR CAAA31484-DW CAAA31485-DM CAAA31485-DR CAAA31485-DW CAAA31486-DM CAAA31486-DR CAAA31486-DW CAAA31487-DM CAAA31487-DR CAAA31487-DW CAAA31488-DM CAAA31488-DR CAAA31488-DT CAAA31488-DW CAAA31489-DM CAAA31489-DR CAAA31489-DW CAAA31490-DM CAAA31490-DR CAAA31490-DT CAAA31490-DW CAAA31490-NP CAAA31492-DM CAAA31492-DR CAAA31492-DW AA31484-QAT AA31485-QAT AA31486-QAT AA31487-QAT AA31488-QAT AA31489-QAT AA31490-QAT AA31492-QAT CAAA31484-QAT CAAA31485-QAT CAAA31486-QAT CAAA31487-QAT CAAA31488-QAT CAAA31489-QAT CAAA31490-QAT CAAA31492-QAT
Ceramic Insulating Tubes, AL23, Both Ends Open, Thin Wall, Thermo Scientific
Tubing Transfer Tubing
Pure Al₂O₃ Aluminium oxide AL23.
  • Corrosion resistant
  • Permeable to microwaves
Dense, excellent thermal and electrical resistance properties.