Dodecylbenzene mixture of isomers

Supplier: Thermo Scientific Chemicals

31362-30 31362-A1
CAAA31362-30EA 241.74 CAD
CAAA31362-30 CAAA31362-A1
Dodecylbenzene mixture of isomers
Dodecylbenzene

CAS No.: 123-01-3 Molecular Formula: C6H5C12H25 Formula Weight: 246.44 Physical Form: Liquid Density: 0.87 Refractive Index: 1.49 Odor: Specific type Melting Point: -3° Boiling Point: 179-180°/13mm MDL No.: MFCD00008974

Formula: C₁₈H₃₀
Boiling Pt: 179…180 °C (13 mmHg)
Melting Pt: –3 °C
Density: 0.87
Storage Temperature: Ambient
MDL Number: MFCD00008974
CAS Number: 123-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR