Nickel on silica-alumina for use as a catalyst

Supplier: Thermo Scientific Chemicals
Danger

31276-06 31276-14 31276-22
CAAA31276-06EA 34.44 CAD
CAAA31276-06 CAAA31276-14 CAAA31276-22
Nickel on silica-alumina for use as a catalyst
Nickel

CAS No.: 7440-02-0 Molecular Formula: 66+/-5% Ni Physical Form: Powder Appearance: Grey Odor: Odorless UN No.: 3178 MDL No.: MFCD00011137

Formula: Ni
MW: 58.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
UN: 3178
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR