Potassium trioxalatoferrate(III) trihydrate

Supplier: Thermo Scientific Chemicals
Warning

31124-14 31124-22
CAAAA31124-14EA 125.29 CAD
CAAAA31124-14 CAAA31124-22
Potassium trioxalatoferrate(III) trihydrate
Potassium trioxalatoferrate(III) trihydrate

CAS No.: 5936-11-8 Molecular Formula: K3Fe(C2O4)3.3H2O Formula Weight: 491.26 (437.21anhy) Physical Form: Crystalline Density: 2.133 Sensitivity: Hygroscopic Appearance: Green UN No.: 3288 MDL No.: MFCD00150450

Formula: K₃Fe(C₂O₄)₃·3H₂O
Density: 2,133
Storage Temperature: Ambient
MDL Number: MFCD00150450
CAS Number: 5936-11-8
EINECS: 238-954-7
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Form Crystalline


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR