Iron(III) oxalate hexahydrate

Supplier: Thermo Scientific Chemicals
Warning

31116-06 31116-14 31116-22
CAAA31116-06EA 85.79 CAD
CAAA31116-06 CAAA31116-14 CAAA31116-22
Iron(III) oxalate hexahydrate
Iron(III) oxalate hexahydrate

CAS No.: 166897-40-1 Molecular Formula: Fe2(C2O4)3.6H2O Formula Weight: 483.84 (375.55anhy) Physical Form: Powder UN No.: 3288 MDL No.: MFCD00054430

Formula: Fe₂(C₂O₄)₃·6H₂O
MW: 483.84 g/mol (375.55 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00054430
CAS Number: 166897-40-1
EINECS: 220-951-7
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR