Cyclopentadienyliron dicarbonyl ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
31114-06
31114-14
31114-22
CAAA31114-06EA
58.46
CAD
CAAA31114-06
CAAA31114-14
CAAA31114-22
Cyclopentadienyliron dicarbonyl ≥99%
Cyclopentadienyliron dicarbonyl
Exists as a dimer
Purity: 99%
CAS No.: 12154-95-9
Molecular Formula: C14H10Fe2O4
Formula Weight: 353.93
Physical Form: Crystalline, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Red
Melting Point: 194° dec.
UN No.: 3466
MDL No.: MFCD00135625
Formula:
C₁₄H₁₂Fe₂O₄ Melting Pt: 194 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00001436 CAS Number: 12154-95-9 EINECS: 235-276-3 UN: 3466 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...