Aluminum metaphosphate (19.4% Al₂O₃, 78.9% P₂O₅)

Supplier: Thermo Scientific Chemicals
Warning

30715-30 30715-A1 30715-D9
CAAA30715-30EA 65.89 CAD
CAAA30715-30 CAAA30715-A1 CAAA30715-D9
Aluminum metaphosphate (19.4% Al₂O₃, 78.9% P₂O₅)
Aluminum metaphosphate

CAS No.: 32823-06-6 Molecular Formula: Al(PO3)3 Formula Weight: 263.90 Physical Form: Powder Density: 2.779 Appearance: White MDL No.: MFCD00075314

Formula: Al(PO₃)₃
Density: 2.779
Storage Temperature: Ambient
MDL Number: MFCD00075314
CAS Number: 32823-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR