Iron(II) ethylenediammonium sulfate tetrahydrate ≥99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ferrous Ethylenediamine Sulphate, Ferrous Ethylenediamine Sulfate
30547-14
30547-22
CAAA30547-14EA
85.64
CAD
CAAA30547-14
CAAA30547-22
Iron(II) ethylenediammonium sulfate tetrahydrate ≥99%
Iron(II) ethylenediammonium sulfate tetrahydrate
Iron(II) ethylenediammonium sulfate tetrahydrate is used in the preparation of gold nanoparticles by reacting with gold(III)-bis[ethylene-diamine]-chloride.
Purity: 99%
CAS No.: 34962-29-3
Molecular Formula: C2H10FeN2O8S2.4H2O
Formula Weight: 382.14 (310.08anhy)
Physical Form: Crystalline
Appearance: White to pale green
MDL No.: MFCD00082474
Formula:
C₂H₈N₂ MW: 310.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00082474 CAS Number: 34962-29-3 EINECS: 264-357-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...