Tetrasodium meso-tetra(sulfonatophenyl)porphine dodecahydrate ≥95%

Supplier: Thermo Scientific Chemicals
Warning

30538-03 30538-MD
CAAA30538-03EA 583.02 CAD
CAAA30538-03 CAAA30538-MD
Tetrasodium meso-tetra(sulfonatophenyl)porphine dodecahydrate ≥95%
Tetrasodium meso-tetra(sulfonatophenyl)porphine dodecahydrate

Purity: 95% CAS No.: 122451-08-5 Molecular Formula: C44H26N4Na4O12S4.12H2O Formula Weight: 1239.11 (1022.87anhy) Physical Form: Crystalline Beilstein Reference: 5488862 MDL No.: MFCD00151011

Formula: C₄₄H₂₆N₄Na₄O₁₂S₄·12H₂O
Storage Temperature: Ambient
MDL Number: MFCD00151011
CAS Number: 122451-08-5
EINECS: 254-262-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR