Bis(2,4-pentanedionato)tin(IV) dichloride ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tin(IV) Acetylacetonate Dichloride, Tin(IV) Chloride Bis(2,4-pentanedionate), Acetylacetone Tin(IV) Dichloride Salt
30532-09
30532-18
CAAA30532-09EA
237
CAD
CAAA30532-09
CAAA30532-18
Bis(2,4-pentanedionato)tin(IV) dichloride ≥95%
Bis(2,4-pentanedionato)tin(IV) dichloride
Purity: 95%
CAS No.: 16919-46-3
Molecular Formula: SnCl2(C5H7O2)2
Formula Weight: 387.83
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Melting Point: 199°
UN No.: 3146
MDL No.: MFCD00015316
Formula:
C₁₀H₁₄Cl₂O₄Sn MW: 387.83 g/mol Melting Pt: 199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015316 CAS Number: 16919-46-3 EINECS: 240-972-5 UN: 3146 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...