Zirconium silicate

Supplier: Thermo Scientific Chemicals
Warning

30477-36 30477-A3 30477-A9
CAAA30477-36EA 76.79 CAD
CAAA30477-36 CAAA30477-A3 CAAA30477-A9
Zirconium silicate
Zirconium silicate

m.p. >1540° dissociates to ZrO2 and SiO2


CAS No.: 10101-52-7 Molecular Formula: ZrSiO4 Formula Weight: 183.30 Physical Form: 2 Micron powder Density: 4.56 Appearance: White Odor: Odorless Melting Point: 2550° MDL No.: MFCD00085353

Formula: O₄SiZr
MW: 183.305 g/mol
Melting Pt: 2550 °C
Density: 4.56
Storage Temperature: Ambient
MDL Number: MFCD00085353
CAS Number: 10101-52-7
EINECS: 233-252-7
Merck Index: 14,10181

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR