Trimethyl thiophosphate
Supplier: Thermo Scientific Chemicals
|
Danger
|
30395-03
30395-06
30395-14
CAAA30395-03EA
43.77
CAD
CAAA30395-03
CAAA30395-06
CAAA30395-14
Trimethyl thiophosphate
Trimethyl thiophosphate
CAS No.: 152-18-1
Molecular Formula: (CH3O)3P(S)
Formula Weight: 156.15
Physical Form: Liquid
Appearance: Off-white
Odor: Sulphurous
UN No.: 3278
MDL No.: MFCD00014888
Formula:
C₃H₉O₃PS Storage Temperature: Ambient |
MDL Number:
MFCD00014888 CAS Number: 152-18-1 UN: 3278 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...