Trimethylphosphine oxide
Supplier: Thermo Scientific Chemicals
|
Warning
|
30394-03
30394-06
CAAA30394-03EA
184.86
CAD
CAAA30394-03
CAAA30394-06
Trimethylphosphine oxide
Trimethylphosphine oxide
CAS No.: 676-96-0
Molecular Formula: (CH3)3P(O)
Formula Weight: 92.08
Physical Form: Crystalline
Odor: Odorless
Melting Point: 140-141°
MDL No.: MFCD00013910
Formula:
C₃H₉OP Melting Pt: 140…141 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013910 CAS Number: 676-96-0 EINECS: 211-633-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...