Triethylphosphine oxide
Supplier: Thermo Scientific Chemicals
|
Warning
|
30391-03
30391-06
CAAA30391-03EA
129.09
CAD
CAAA30391-03
CAAA30391-06
Triethylphosphine oxide
Triethylphosphine oxide
CAS No.: 597-50-2
Molecular Formula: (C2H5)3P(O)
Formula Weight: 134.16
Physical Form: Crystalline
Appearance: White
Melting Point: 48-50°
MDL No.: MFCD00013914
Formula:
C₆H₁₅OP Melting Pt: 48…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013914 CAS Number: 597-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...