Tricyclohexylphosphine oxide
Supplier: Thermo Scientific Chemicals
|
Danger
|
30387-03
30387-06
30387-14
CAAA30387-03EA
70.47
CAD
CAAA30387-03
CAAA30387-06
CAAA30387-14
Tricyclohexylphosphine oxide
Tricyclohexylphosphine oxide
CAS No.: 13689-19-5
Molecular Formula: (C6H11)3P(O)
Formula Weight: 296.42
Physical Form: Crystalline
Melting Point: 155-157°
UN No.: 3464
MDL No.: MFCD00014298
Formula:
C₁₈H₃₃OP Melting Pt: 155…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014298 CAS Number: 13689-19-5 UN: 3464 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...