1,3-Propylenediphosphonic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
30277-02
30277-04
30277-09
CAAA30277-02EA
48.51
CAD
CAAA30277-02
CAAA30277-04
CAAA30277-09
1,3-Propylenediphosphonic acid 98+%
1,3-Propylenediphosphonic acid
Purity: 98+%
CAS No.: 4671-82-3
Molecular Formula: [P(O)(OH)2CH2]2CH2
Formula Weight: 204.06
Physical Form: Powder
Odor: Odorless
MDL No.: MFCD00050066
Formula:
C₃H₁₀O₆P₂ Melting Pt: 178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00050066 CAS Number: 4671-82-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...