Di-tert-butyl phosphonate ≥95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Di-tert-butyl phosphite

30233-03 30233-06 30233-14
CAAA30233-03EA 62.88 CAD
CAAA30233-03 CAAA30233-06 CAAA30233-14
Di-tert-butyl phosphonate ≥95%
Di-tert-butyl phosphonate

Applications: solvent, as an antioxidant, and as an intermediate
Soluble in organic solvents


Purity: 95% CAS No.: 13086-84-5 Molecular Formula: C8H19O3P Formula Weight: 194.21 Physical Form: Liquid Density: 0.995 Refractive Index: 1.4200 Sensitivity: Air and Moisture Sensitive Boiling Point: 66-68°/0.5mm Solubility: Soluble in organic solvents MDL No.: MFCD00014999


As a solvent, as an antioxidant, and as an intermediate

Formula: C₈H₁₉O₃P
MW: 194.21 g/mol
Boiling Pt: 66…68 °C (0.5 mmHg)
Density: 0.995
Storage Temperature: Ambient
MDL Number: MFCD00995019
CAS Number: 13086-84-5
EINECS: 235-996-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR