Tris(2-cyanoethyl)phosphine ≥94%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tris (2-cyanoethyl)phosphine, 3,3',3''-Phosphinetriyltris(propiononitrile), 3,3',3''-Phosphinidynetris(propiononitrile)
30165-03
30165-09
30165-18
CAAA30165-03EA
53.88
CAD
CAAA30165-03
CAAA30165-09
CAAA30165-18
Tris(2-cyanoethyl)phosphine ≥94%
Tris(2-cyanoethyl)phosphine
Purity: 94%
CAS No.: 4023-53-4
Molecular Formula: (NCCH2CH2)3P
Formula Weight: 193.19
Physical Form: Crystalline
Sensitivity: Air and Moisture Sensitive
Melting Point: 97-98°
Boiling Point: 235°/0.9mm
UN No.: 3464
MDL No.: MFCD00013828
Formula:
C₉H₁₂N₃P MW: 193.19 g/mol Boiling Pt: 235 °C (0.9 mmHg) Melting Pt: 97…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013828 CAS Number: 4023-53-4 EINECS: 223-687-0 UN: 3464 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...