Ammonium O,O'-diethyl dithiophosphate ≥95%

Supplier: Thermo Scientific Chemicals

30139-06 30139-14
CAAA30139-06EA 52.46 CAD
CAAA30139-06 CAAA30139-14
Ammonium O,O'-diethyl dithiophosphate ≥95%
Ammonium O,O'-diethyl dithiophosphate

Purity: 95% CAS No.: 1068-22-0 Molecular Formula: C4H14NO2PS2 Formula Weight: 203.25 Physical Form: Crystalline Appearance: White Odor: Unpleasant Melting Point: 165-175° MDL No.: MFCD00012635

Formula: C₄H₁₄NO₂PS₂
Melting Pt: 165…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00012635
CAS Number: 1068-22-0
EINECS: 213-942-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR