Calcium tetrafluoroborate hydrate

Supplier: Thermo Scientific Chemicals
Danger

30131-14 30131-22
CAAA30131-14EA 144.25 CAD
CAAA30131-14 CAAA30131-22
Calcium tetrafluoroborate hydrate
Calcium tetrafluoroborate hydrate

CAS No.: 15978-68-4 Molecular Formula: Ca(BF4)2.xH2O Formula Weight: 213.68(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00015972

Formula: Ca(BF₄)₂·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00015972
CAS Number: 13814-93-2
EINECS: 237-485-5
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR