Ceramic Tubes, AL24, One End Closed, Thermo Scientific
Supplier: Thermo Scientific Chemicals
30093-DR 30093-DW
CAAA30093-DMEA 283.07 CAD
CAAA30093-DM CAAA30093-DR CAAA30093-DW CAAA30093-EH CAAA30093-EM CAAA30093-EO AA30093-QAT CAAA30093-QAT
Ceramic Tubes, AL24, One End Closed, Thermo Scientific
Tubing Transfer Tubing
Pure Al₂O₃ Aluminium oxide AL24.
  • Slightly porous
  • Good resistance to thermal shock
Extremely good creep strength.