Ceramic Tubes, AL23, One End Closed, Thermo Scientific
Supplier: Thermo Scientific Chemicals
30076-DM 30076-DW 30078-DM 30078-DR 30078-DW 30083-EM
CAAA30076-DMEA 200.66 CAD
CAAA30076-DM CAAA30076-DR CAAA30076-DW CAAA30076-EH CAAA30076-EM CAAA30076-EO CAAA30077-DM CAAA30077-DR CAAA30077-DW CAAA30077-EH CAAA30077-EM CAAA30077-EO CAAA30078-DM CAAA30078-DR CAAA30078-DW CAAA30078-EH CAAA30078-EM CAAA30078-EO CAAA30080-DM CAAA30080-DR CAAA30080-DW CAAA30080-EH CAAA30080-EM CAAA30083-DM CAAA30083-DR CAAA30083-DW CAAA30083-EH CAAA30083-EM CAAA30084-DM CAAA30084-DR CAAA30084-DW CAAA30084-EH CAAA30084-EM CAAA30086-DM CAAA30086-DR CAAA30086-DW CAAA30086-EH CAAA30086-EM CAAA30088-DM CAAA30088-DR CAAA30088-DW CAAA30088-EH CAAA30088-EM AA30076-QAT AA30077-QAT AA30078-QAT AA30080-QAT AA30083-QAT AA30084-QAT AA30086-QAT AA30088-QAT CAAA30076-QAT CAAA30077-QAT CAAA30078-QAT CAAA30080-QAT CAAA30083-QAT CAAA30084-QAT CAAA30086-QAT CAAA30088-QAT
Ceramic Tubes, AL23, One End Closed, Thermo Scientific
Tubing Transfer Tubing
Pure Al₂O₃ Aluminium oxide AL23.
  • Corrosion resistant
  • Permeable to microwaves
Dense, excellent thermal and electrical resistance properties.
Outer diameter ranges from 16 to 70 mm.
Order Now
SPECIFICATIONS
Environmentally Preferable