Copper(II) tetrafluoroborate hexahydrate ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
26127-22
26127-36
CAAA26127-22EA
143.62
CAD
CAAA26127-22
CAAA26127-36
Copper(II) tetrafluoroborate hexahydrate ≥98%
Copper(II) tetrafluoroborate hexahydrate
Purity: 98%
CAS No.: 72259-10-0
Molecular Formula: Cu(BF4)2.6H2O
Formula Weight: 345.25 (237.15anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
UN No.: 3260
MDL No.: MFCD00150223
Formula:
B₂CuF₈H₁₂O₆ MW: 345.243 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150223 CAS Number: 72259-10-0 EINECS: 253-959-4 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...