Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate
Supplier: Thermo Scientific Chemicals
|
Warning
|
24127-03
24127-06
24127-14
24127-22
CAAA24127-03EA
50.88
CAD
CAAA24127-03
CAAA24127-06
CAAA24127-14
CAAA24127-22
Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate
Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate
CAS No.: 16165-32-5
Molecular Formula: C6H24Cl3CrN6.3.5H2O
Formula Weight: 401.71 (338.65anhy)
Physical Form: Crystalline
Appearance: Yellow to orange
MDL No.: MFCD00150160
Formula:
C₆H₂₄Cl₃CrN₆.3,5H₂O MW: 401.71 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150160 CAS Number: 16165-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...