Chromium(III) phosphate tetrahydrate

Supplier: Thermo Scientific Chemicals
Warning

24110-09 24110-18
CAAA24110-09EA 71.42 CAD
CAAA24110-09 CAAA24110-18
Chromium(III) phosphate tetrahydrate
Chromium(III) phosphate tetrahydrate

Applications: Catalyst, pigment


CAS No.: 10101-59-4 Molecular Formula: CrPO4.4H2O Formula Weight: 219.04 (146.97anhy) Physical Form: Powder Density: 2.26 MDL No.: MFCD00016049


Applications: Catalyst, pigment

Formula: CrH₈O₈P
Density: 2,26
Storage Temperature: Ambient
MDL Number: MFCD00016049
CAS Number: 10101-59-4
EINECS: 232-141-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR