Tricarbonylnitrosylcobalt
Supplier: Thermo Scientific Chemicals
|
Danger
|
23151-03
23151-06
CAAA23151-03EA
125.77
CAD
CAAA23151-03
CAAA23151-06
Tricarbonylnitrosylcobalt
Tricarbonylnitrosylcobalt
CAS No.: 14096-82-3
Molecular Formula: C3CoNO4
Formula Weight: 172.97
Physical Form: Liquid
Density: 1.47
Sensitivity: Air Sensitive
Appearance: Dark red
Odor: Unpleasant
Melting Point: -1°
Boiling Point: 48.6°(55° dec.)
UN No.: 1992
MDL No.: MFCD00016014
Formula:
C₃CoNO₄ Boiling Pt: 48.6 °C/ 55 °C (boil./ dec.) Melting Pt: –1 °C Density: 1,47 Storage Temperature: Ambient |
MDL Number:
MFCD00016014 CAS Number: 14096-82-3 EINECS: 237-945-5 UN: 1992 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...