Cobalt(II) tetrafluoroborate hydrate ≥96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
23142-14
23142-22
CAAA23142-14EA
94.8
CAD
CAAA23142-14
CAAA23142-22
Cobalt(II) tetrafluoroborate hydrate ≥96%
Cobalt(II) tetrafluoroborate hydrate
Purity: 96%
CAS No.: 26490-63-1
Molecular Formula: Co(BF4)2.xH2O
Formula Weight: 232.54(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
UN No.: 3260
MDL No.: MFCD00016012
Formula:
Co(BF₄)₂.xH₂O MW: 232.54 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00016012 CAS Number: 26490-63-1 UN: 3260 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...