Dicarbonylcyclopentadienylcobalt

Supplier: Thermo Scientific Chemicals
Danger

23136-03 23136-06
CAAA23136-03EA 81.53 CAD
CAAA23136-03 CAAA23136-06
Dicarbonylcyclopentadienylcobalt
Dicarbonylcyclopentadienylcobalt

Applications: Catalyst for the cyclotrimerization of alkynes


CAS No.: 12078-25-0 Molecular Formula: C7H5CoO2 Formula Weight: 180.05 Flash Point: 26°(80°F) Physical Form: Liquid Sensitivity: Air Sensitive Boiling Point: 139-140°/710mm UN No.: 3281 MDL No.: MFCD00009589


Applications: Catalyst for the cyclotrimerization of alkynes

Formula: C₇H₅CoO₂
Boiling Pt: 139…140 °C (710 mmHg)
Flash Pt: 26 °C (80 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009589
CAS Number: 12078-25-0
EINECS: 235-139-8
UN: 3281
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR