Potassium hexacyanocobaltate(III) ≥90%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tripotassium hexacyanocobaltate(III)
23126-18
23126-30
CAAA23126-18EA
164.32
CAD
CAAA23126-18
CAAA23126-30
Potassium hexacyanocobaltate(III) ≥90%
Potassium hexacyanocobaltate(III)
Soluble in water and acetic acid solutions. Insoluble in alcohol
Purity: 90+%
CAS No.: 13963-58-1
Molecular Formula: C6CoK3N6
Formula Weight: 332.33
Physical Form: Crystalline
Density: 1.906
Appearance: Pale yellow to pale brown
Odor: Odorless
Melting Point: dec.
Solubility: Soluble in water and acetic acid solutions. Insoluble in alcohol
MDL No.: MFCD00011391
Formula:
C₆CoK₃N₆ Melting Pt: Dec. Density: 1.906 Storage Temperature: Ambient |
MDL Number:
MFCD00011391 CAS Number: 13963-58-1 EINECS: 237-742-1 Merck Index: 14,7637 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...