Cobalt(II) phosphate octahydrate
Supplier: Thermo Scientific Chemicals
|
Danger
|
23111-18
23111-30
CAAA23111-18EA
202.24
CAD
CAAA23111-18
CAAA23111-30
Cobalt(II) phosphate octahydrate
Cobalt(II) phosphate octahydrate
CAS No.: 10294-50-5
Molecular Formula: Co3(PO4)2.8H2O
Formula Weight: 510.87 (366.74anhy)
Physical Form: Powder
Density: 2.769
Odor: Odorless
MDL No.: MFCD00016028
Formula:
Co₃(PO₄)₂·8H₂O Density: 2.769 Storage Temperature: Ambient |
MDL Number:
MFCD00016028 CAS Number: 10294-50-5 EINECS: 236-655-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...