Titanium trialuminide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

22895-14 22895-22
CAAA22895-14EA 243.32 CAD
CAAA22895-14 CAAA22895-22
Titanium trialuminide ≥99.5% (metals basis)
Titanium trialuminide

Purity: 99.5% CAS No.: 12004-78-3 Molecular Formula: TiAl3 Physical Form: -325 Mesh Powder, Intermetallic Odor: Odorless UN No.: 3178 MDL No.: MFCD00054094

Formula: Al₃Ti
Storage Temperature: Ambient
MDL Number: MFCD00054094
CAS Number: 12004-78-3
EINECS: 234-461-6
UN: 3178
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR