n-Butyl phosphate mixture of mono-n-butyl and di-n-butyl
Supplier: Thermo Scientific Chemicals
|
Danger
|
22404-30
22404-A1
CAAA22404-30EA
75.84
CAD
CAAA22404-30
CAAA22404-A1
n-Butyl phosphate mixture of mono-n-butyl and di-n-butyl
n-Butyl phosphate
Approximately 1:1 molar ratio of alkyl & dialkyl forms. Decomposition occurs above 125°
CAS No.: 52933-01-4
Molecular Formula: [CH3(CH2)3O]2P(O)OH/[CH3(CH2)3O]P(O)(OH)2
Flash Point: 110°(230°F)
Physical Form: Liquid
Density: 1.123
Refractive Index: 1.4290
Appearance: Colorless
Melting Point: -60°
Boiling Point:
Formula:
[CH₃(CH₂)₃O]₂P(O)OH/[CH₃(CH₂)₃O]P(O)(OH)₂ Boiling Pt: >124 °C (dec.) Melting Pt: –60 °C Density: 1,123 Flash Pt: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013947 CAS Number: 52933-01-4 UN: 1718 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...