Cerium(III) oxalate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
21122-14
21122-22
CAAA21122-06EA
121.5
CAD
CAAA21122-06
CAAA21122-14
CAAA21122-22
CAAA21122-36
Cerium(III) oxalate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Cerium (III) oxalate hydrate
Purity: 99.9%
CAS No.: 15750-47-7
Molecular Formula: Ce2(C2O4)3.xH2O
Formula Weight: 544.29(anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Merck Reference: 14,2001
UN No.: 3288
MDL No.: MFCD00150139
Formula:
C₆Ce₂O₁₂·H₂O Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00150139 CAS Number: 15750-47-7 UN: 3288 ADR: 6.1,III Merck Index: 14,2001 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...