Cerium(IV) oxide, hydrated
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cerium(IV) hydroxide
21120-18
21120-30
CAAA21120-18EA
90.69
CAD
CAAA21120-18
CAAA21120-30
Cerium(IV) oxide, hydrated
Cerium(IV) oxide, hydrated
CAS No.: 12014-56-1
Molecular Formula: CeO2.xH2O
Formula Weight: 172.12(anhy)
Physical Form: Lump/Powder
Appearance: Yellow to pale brown
Odor: Odorless
UN No.: 3262
MDL No.: MFCD00010927
Formula:
CeO₂.xH₂O MW: 208.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010927 CAS Number: 12014-56-1 EINECS: 234-599-7 UN: 3262 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...