Cerium(III) acetylacetonate hydrate
Supplier: Thermo Scientific Chemicals
|
Warning
|
21116-22
21116-36
CAAA21116-22EA
314.42
CAD
CAAA21116-22
CAAA21116-36
Cerium(III) acetylacetonate hydrate
Cerium(III) acetylacetonate hydrate
CAS No.: 206996-61-4
Molecular Formula: C15H21CeO6.xH2O
Formula Weight: 437.44(anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Yellow to red
Melting Point: 131-132°
MDL No.: MFCD00150164
Formula:
C₁₅H₂₁CeO₆•xH₂O Melting Pt: 131…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150164 CAS Number: 206996-61-4 EINECS: 239-720-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...