Silver lactate
Supplier: Thermo Scientific Chemicals
|
Warning
|
20835-09
20835-18
CAAA20835-09EA
203.82
CAD
CAAA20835-09
CAAA20835-18
Silver lactate
Silver lactate
CAS No.: 128-00-7
Molecular Formula: C3H5AgO3
Formula Weight: 196.94
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White to grey
Odor: Odorless
Melting Point: 120-122°
UN No.: 3077
MDL No.: MFCD00043279
Formula:
C₃H₅AgO₃ Melting Pt: 120…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043279 CAS Number: 15768-18-0 UN: 3077 ADR: 9,III Merck Index: 14,8517 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...