Methyl malonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl 3-chloro-3-oxopropanoate

20823-14 20823-06
CAAAA20823-14EA 312.84 CAD
CAAAA20823-14 CAAA20823-03 CAAA20823-06
Methyl malonyl chloride 97%
Methyl malonyl chloride

Purity: 97% CAS No.: 37517-81-0 Molecular Formula: C4H5ClO3 Formula Weight: 136.53 Flash Point: 80°(176°F) Physical Form: Liquid Density: 1.27 Refractive Index: 1.4325 Sensitivity: Moisture Sensitive Appearance: Colorless Boiling Point: 58°/12mm UN No.: 3265 MDL No.: MFCD00013657

Formula: C₄H₅ClO₃
MW: 136.53 g/mol
Boiling Pt: 58 °C (12 mmHg)
Density: 1.27
Flash Pt: 80 °C (176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013657
CAS Number: 37517-81-0
EINECS: 253-540-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR