2,6-Xylyl isocyanide
Supplier: Thermo Scientific Chemicals
|
Danger
|
20748-03
20748-06
CAAA20748-03EA
143.78
CAD
CAAA20748-03
CAAA20748-06
2,6-Xylyl isocyanide
2,6-Xylyl isocyanide
CAS No.: 2769-71-3
Molecular Formula: C9H9N
Formula Weight: 131.18
Physical Form: Crystalline
Odor: Unpleasant
Melting Point: 73-74°
UN No.: 2811
MDL No.: MFCD00013479
Formula:
C₉H₉N Melting Pt: 73…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013479 CAS Number: 2769-71-3 EINECS: 220-459-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...